Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300022324.mm.1 |
NM_001190156.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022324.mm.1 |
NM_001190156.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022324.mm.1 |
NM_029655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022324.mm.1 |
NM_029655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022324.mm.1 |
TC0300002822.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022322.mm.1 |
NM_001190156.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022322.mm.1 |
NM_001190156.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022322.mm.1 |
NM_029655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022322.mm.1 |
NM_029655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022322.mm.1 |
TC0300002822.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022325.mm.1 |
NM_001190156.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022325.mm.1 |
NM_001190156.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022325.mm.1 |
NM_029655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022325.mm.1 |
NM_029655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022325.mm.1 |
TC0300002822.mm.1 |
MTA TC |