Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sucla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011506.3 | NP_035636.1 | 2 - 3 | 292 | 73 | |
GenBank mRNA | AF058955.1 | - | 1 - 2 | 159 | 73 | |
AK005273.1 | - | 2 - 3 | 274 | 73 | ||
AK081965.1 | - | 2 - 3 | 313 | 73 | ||
AK088801.1 | - | 2 - 3 | 293 | 73 | ||
AK132762.1 | - | 2 - 3 | 282 | 73 | ||
AK150594.1 | - | 2 - 3 | 292 | 73 | ||
AK152783.1 | - | 2 - 3 | 292 | 73 | ||
AK153198.1 | - | 2 - 3 | 292 | 73 | ||
AK160130.1 | - | 2 - 3 | 293 | 73 | ||
AK160652.1 | - | 2 - 3 | 281 | 73 | ||
BC056353.1 | - | 2 - 3 | 283 | 6808360 | 73 | |
BC057605.1 | - | 2 - 3 | 283 | 6808360 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |