Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026816.3 | NP_081092.1 | 1 - 2 | 183 | 76 | |
GenBank mRNA | AK144858.1 | - | 1 - 2 | 184 | 76 | |
BC027173.1 | - | 1 - 2 | 179 | 3986466 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |