Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027581.1 | NP_081857.1 | 6 - 7 | 1370 | 156 | |
GenBank mRNA | AK014850.1 | - | 5 - 6 | 1102 | 156 | |
BC145796.1 | - | 6 - 7 | 1365 | 40131099 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |