Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptrhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204912.1 | NP_001191841.1 | 1 - 2 | 249 | 53 | |
GenBank mRNA | AK010531.1 | - | 1 - 2 | 173 | 53 | |
BC067010.1 | - | 1 - 2 | 213 | 53 | ||
BY339231.1 | - | 1 - 2 | 234 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |