Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162979.1 | NP_001156451.1 | 10 - 11 | 1592 | 130 | |
GenBank mRNA | AK014876.1 | - | 10 - 11 | 1575 | 130 | |
AK029544.1 | - | 10 - 11 | 1591 | 130 | ||
BC127049.1 | - | 10 - 11 | 1579 | 130 | ||
BC152307.1 | - | 10 - 11 | 1581 | 40062409 | 130 | |
BC152308.1 | - | 10 - 11 | 1581 | 40062416 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |