Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133815.2 | NP_598576.2 | 7 - 8 | 1056 | 89 | |
GenBank mRNA | AK028426.1 | - | 7 - 8 | 1033 | 89 | |
AK030606.1 | - | 7 - 8 | 1010 | 89 | ||
AK033459.1 | - | 7 - 8 | 1008 | 89 | ||
AK088179.1 | - | 7 - 8 | 1009 | 89 | ||
AK151798.1 | - | 7 - 8 | 1023 | 89 | ||
AK161762.1 | - | 7 - 8 | 1034 | 89 | ||
AK166850.1 | - | 7 - 8 | 996 | 89 | ||
AK167157.1 | - | 7 - 8 | 1001 | 89 | ||
BC021516.1 | - | 7 - 8 | 997 | 5352337 | 89 | |
BC025583.1 | - | 6 - 7 | 806 | 89 | ||
BC042522.1 | - | 7 - 8 | 1016 | 4216442 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |