Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010022.3 | NP_034152.2 | 5 - 6 | 624 | 113 | |
GenBank mRNA | AK075831.1 | - | 5 - 6 | 606 | 113 | |
AK165959.1 | - | 5 - 6 | 634 | 113 | ||
BC055890.1 | - | 5 - 6 | 580 | 4242586 | 113 | |
L42996.1 | - | 5 - 6 | 577 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |