Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010022.3 | NP_034152.2 | 7 - 8 | 1006 | 76 | |
GenBank mRNA | AK075831.1 | - | 7 - 8 | 988 | 76 | |
AK165959.1 | - | 7 - 8 | 1016 | 76 | ||
BC055890.1 | - | 7 - 8 | 962 | 4242586 | 76 | |
L42996.1 | - | 7 - 8 | 959 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |