Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164493.1 | NP_001157965.1 | 3 - 4 | 1019 | 112 | |
GenBank mRNA | AK122572.1 | - | 3 - 4 | 672 | 112 | |
BC065168.1 | - | 3 - 4 | 1019 | 112 | ||
BC138283.1 | - | 2 - 3 | 375 | 8861302 | 112 | |
BC145230.1 | - | 2 - 3 | 375 | 112 | ||
BC145748.1 | - | 2 - 3 | 375 | 40131051 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |