Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sass6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289568.1 | NP_001276497.1 | 9 - 10 | 1105 | 116 | |
NM_001289571.1 | NP_001276500.1 | 7 - 8 | 961 | 116 | ||
NM_028349.3 | NP_082625.2 | 8 - 9 | 1022 | 116 | ||
GenBank mRNA | AK013338.1 | - | 9 - 10 | 997 | 116 | |
AK145076.1 | - | 7 - 8 | 910 | 116 | ||
AK165224.1 | - | 9 - 10 | 1051 | 116 | ||
BC050110.1 | - | 8 - 9 | 972 | 30024366 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |