Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289524.1 | NP_001276453.1 | 12 - 13 | 1470 | 86 | |
NM_001289525.1 | NP_001276454.1 | 10 - 11 | 1217 | 86 | ||
NM_024194.6 | NP_077156.2 | 12 - 13 | 1473 | 86 | ||
GenBank mRNA | AK011726.1 | - | 12 - 13 | 1454 | 86 | |
AK017814.1 | - | 12 - 13 | 1442 | 86 | ||
AK034978.1 | - | 1453 | 86 | |||
AK148748.1 | - | 12 - 13 | 1433 | 86 | ||
AK152244.1 | - | 10 - 11 | 1198 | 86 | ||
AK163288.1 | - | 12 - 13 | 1441 | 86 | ||
AK170052.1 | - | 12 - 13 | 1450 | 86 | ||
BC002279.1 | - | 8 - 9 | 862 | 3497400 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |