Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103182.2 | NP_001096652.2 | 12 - 13 | 1461 | 108 | |
NR_120498.1 | 12 - 13 | 1459 | 108 | |||
GenBank mRNA | AF190325.1 | - | 12 - 13 | 1277 | 108 | |
AK043334.1 | - | 1 - 2 | 1797 | 108 | ||
BC043691.1 | - | 1 - 2 | 338 | 108 | ||
BC069181.1 | - | 9 - 10 | 1000 | 108 | ||
BC088992.1 | - | 12 - 13 | 1249 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |