Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026083.2 | NP_080359.2 | 17 - 18 | 5158 | 101 | |
GenBank mRNA | BC023707.1 | - | 4 - 5 | 1026 | 101 | |
BC082576.1 | - | 6 - 7 | 2670 | 101 | ||
BC094575.1 | - | 10 - 11 | 3707 | 101 | ||
BC138264.1 | - | 17 - 18 | 5110 | 8861283 | 101 | |
BG062845.2 | - | 1 - 2 | 174 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |