Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyw3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168358.1 | NP_001161830.1 | 3 - 4 | 427 | 70 | |
NM_172474.5 | NP_766062.1 | 3 - 4 | 427 | 70 | ||
GenBank mRNA | AK030344.1 | - | 3 - 4 | 425 | 70 | |
AK034820.1 | - | 3 - 4 | 412 | 70 | ||
BC086686.1 | - | 3 - 4 | 401 | 30680486 | 70 | |
BC087879.1 | - | 3 - 4 | 391 | 6533208 | 70 | |
BC141040.1 | - | 3 - 4 | 406 | 9055652 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |