Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159672.1 | NP_001153144.1 | 11 - 12 | 1242 | 85 | |
NM_175317.3 | NP_780526.2 | 11 - 12 | 1271 | 85 | ||
GenBank mRNA | AK028676.1 | - | 11 - 12 | 1244 | 85 | |
AK031647.1 | - | 11 - 12 | 1228 | 85 | ||
AK049698.1 | - | 11 - 12 | 1230 | 85 | ||
BC031852.1 | - | 5 - 6 | 517 | 85 | ||
BC045616.1 | - | 11 - 12 | 1225 | 4983234 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |