Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177893.3 | NP_808561.2 | 7 - 8 | 2474 | 102 | |
GenBank mRNA | AK173087.1 | - | 6 - 7 | 1932 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |