Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrch1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033439.3 | NP_001028611.2 | 10 - 11 | 1492 | 84 | |
NM_001252132.1 | NP_001239061.1 | 10 - 11 | 1492 | 84 | ||
GenBank mRNA | AK129264.1 | - | 10 - 11 | 1481 | 84 | |
AK136026.1 | - | 10 - 11 | 1098 | 84 | ||
AK143909.1 | - | 10 - 11 | 1491 | 84 | ||
AK160902.1 | - | 8 - 9 | 868 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |