Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026083.2 | NP_080359.2 | 10 - 11 | 2206 | 85 | |
GenBank mRNA | AK014200.1 | - | 10 - 11 | 1987 | 85 | |
AK078225.1 | - | 10 - 11 | 2197 | 85 | ||
BC038882.1 | - | 3 - 4 | 1016 | 85 | ||
BC094575.1 | - | 2 - 3 | 810 | 85 | ||
BC138264.1 | - | 10 - 11 | 2155 | 8861283 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |