Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027617.3 | NP_081893.1 | 10 - 11 | 1188 | 157 | |
GenBank mRNA | BC139279.1 | - | 9 - 10 | 1101 | 8862301 | 157 |
BC139280.1 | - | 9 - 10 | 1101 | 8862302 | 157 | |
BC171970.1 | - | 9 - 10 | 1099 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |