Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293561.1 | NP_001280490.1 | 9 - 10 | 1513 | 148 | |
NM_009472.4 | NP_033498.1 | 8 - 9 | 1456 | 148 | ||
GenBank mRNA | AK031655.1 | - | 9 - 10 | 1480 | 148 | |
AK141181.1 | - | 9 - 10 | 1479 | 148 | ||
AK142411.1 | - | 9 - 10 | 1479 | 148 | ||
AK142974.1 | - | 9 - 10 | 1407 | 148 | ||
AK164239.1 | - | 8 - 9 | 1257 | 148 | ||
BC115772.1 | - | 9 - 10 | 1503 | 40085998 | 148 | |
U72634.1 | - | 8 - 9 | 1456 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |