Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnldc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034866.1 | NP_001030038.1 | 11 - 12 | 1022 | 64 | |
GenBank mRNA | BC158033.1 | - | 11 - 12 | 1022 | 9007336 | 64 |
BC172002.1 | - | 11 - 12 | 1025 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |