Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700000941.mm.1 |
NM_026644.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000941.mm.1 |
NM_026644.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000941.mm.1 |
TC1700000117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000943.mm.1 |
NM_026644.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000943.mm.1 |
NM_026644.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000943.mm.1 |
TC1700000117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000944.mm.1 |
NM_026644.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000944.mm.1 |
NM_026644.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000944.mm.1 |
TC1700000117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000945.mm.1 |
NM_026644.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000945.mm.1 |
NM_026644.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000945.mm.1 |
TC1700000117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000946.mm.1 |
NM_026644.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000946.mm.1 |
NM_026644.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000946.mm.1 |
TC1700000117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700000947.mm.1 |
NM_026644.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000947.mm.1 |
NM_026644.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700000947.mm.1 |
TC1700000117.mm.1 |
MTA TC |