Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253712.1 | NP_001240641.1 | 2 - 3 | 404 | 90 | |
NM_001253716.1 | NP_001240645.1 | 2 - 3 | 132 | 90 | ||
NM_026322.4 | NP_080598.2 | 7 - 8 | 803 | 90 | ||
GenBank mRNA | AK002356.1 | - | 2 - 3 | 279 | 90 | |
AK009822.1 | - | 2 - 3 | 281 | 90 | ||
AK018338.1 | - | 2 - 3 | 126 | 90 | ||
AK030278.1 | - | 2 - 3 | 282 | 90 | ||
AK049714.1 | - | 2 - 3 | 405 | 90 | ||
BB652761.1 | - | 2 - 3 | 402 | 90 | ||
BC089311.1 | - | 7 - 8 | 803 | 6412625 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |