Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081063.1 | NP_001074532.1 | 3 - 4 | 527 | 75 | |
GenBank mRNA | AK016601.1 | - | 2 - 3 | 561 | 75 | |
BC115562.2 | - | 3 - 4 | 528 | 40101914 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |