Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027436.3 | NP_081712.2 | 15 - 16 | 1841 | 58 | |
NR_040642.1 | 15 - 16 | 1841 | 58 | |||
GenBank mRNA | AK017491.1 | - | 15 - 16 | 1817 | 58 | |
AK038479.1 | - | 15 - 16 | 1835 | 58 | ||
AK076613.1 | - | 15 - 16 | 1771 | 58 | ||
AK146428.1 | - | 16 - 17 | 1832 | 58 | ||
BC049871.1 | - | 6 - 7 | 681 | 58 | ||
BC110358.1 | - | 15 - 16 | 1819 | 5322591 | 58 | |
BC144798.1 | - | 15 - 16 | 1822 | 58 | ||
BC145711.1 | - | 15 - 16 | 1822 | 40131014 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |