Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009618.2 | NP_033748.2 | 16 - 17 | 1823 | 86 | |
GenBank mRNA | AK016550.1 | - | 16 - 17 | 1823 | 86 | |
AK161266.1 | - | 16 - 17 | 1812 | 86 | ||
U16242.1 | - | 16 - 17 | 1812 | 86 | ||
U22057.1 | - | 15 - 16 | 1763 | 86 | ||
U38806.1 | - | 16 - 17 | 1828 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mus musculus Probe spans exons |