Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400015687.mm.1 |
NM_001164171.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015687.mm.1 |
NM_001164171.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015687.mm.1 |
NM_010856.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015687.mm.1 |
NM_010856.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015687.mm.1 |
TC1400002073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015688.mm.1 |
NM_001164171.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015688.mm.1 |
NM_001164171.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015688.mm.1 |
NM_010856.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015688.mm.1 |
NM_010856.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015688.mm.1 |
TC1400002073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015690.mm.1 |
NM_001164171.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015690.mm.1 |
NM_001164171.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015690.mm.1 |
NM_010856.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015690.mm.1 |
NM_010856.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015690.mm.1 |
TC1400002073.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015689.mm.1 |
NM_001164171.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015689.mm.1 |
NM_001164171.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015689.mm.1 |
NM_010856.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015689.mm.1 |
NM_010856.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015689.mm.1 |
TC1400002073.mm.1 |
MTA TC |