Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epsti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029495.2 | NP_083771.1 | 2 - 3 | 486 | 59 | |
NM_178825.4 | NP_849147.1 | 2 - 3 | 486 | 59 | ||
GenBank mRNA | AK017174.1 | - | 2 - 3 | 319 | 59 | |
AK077223.1 | - | 2 - 3 | 292 | 59 | ||
AK077376.1 | - | 2 - 3 | 321 | 59 | ||
AK135710.1 | - | 2 - 3 | 480 | 59 | ||
AK136055.1 | - | 2 - 3 | 487 | 59 | ||
AY239223.1 | - | 2 - 3 | 319 | 59 | ||
AY239224.1 | - | 2 - 3 | 392 | 59 | ||
BC020120.1 | - | 2 - 3 | 319 | 3988503 | 59 | |
BC021821.1 | - | 2 - 3 | 346 | 3581940 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |