Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd11b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008288.2 | NP_032314.2 | 1 - 2 | 103 | 91 | |
GenBank mRNA | AI182353.1 | - | 1 - 2 | 103 | 91 | |
BC132364.1 | - | 1 - 2 | 43 | 40130641 | 91 | |
BC138780.1 | - | 1 - 2 | 43 | 8861802 | 91 | |
BC145415.1 | - | 1 - 2 | 43 | 9053973 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |