Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diexf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145415.2 | NP_663390.2 | 5 - 6 | 793 | 141 | |
GenBank mRNA | AK052355.1 | - | 5 - 6 | 794 | 141 | |
AK088756.1 | - | 5 - 6 | 806 | 141 | ||
AK147251.1 | - | 5 - 6 | 787 | 141 | ||
BC019218.1 | - | 6 - 7 | 753 | 5065857 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |