Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traf3ip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153137.4 | NP_694777.3 | 5 - 6 | 967 | 92 | |
GenBank mRNA | AK031400.1 | - | 5 - 6 | 907 | 92 | |
AK041994.1 | - | 5 - 6 | 968 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |