Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199020.1 | NP_001185949.1 | 3 - 4 | 439 | 142 | |
NM_021421.4 | NP_067396.3 | 2 - 3 | 574 | 142 | ||
NR_037578.1 | 2 - 3 | 533 | 142 | |||
GenBank mRNA | AK012003.1 | - | 2 - 3 | 570 | 142 | |
AK017545.1 | - | 2 - 3 | 537 | 142 | ||
AK077039.1 | - | 2 - 3 | 520 | 142 | ||
AK135709.1 | - | 3 - 4 | 436 | 142 | ||
AK158731.1 | - | 2 - 3 | 590 | 142 | ||
AK166619.1 | - | 3 - 4 | 438 | 142 | ||
BC024907.1 | - | 2 - 3 | 534 | 2812124 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |