Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205075.1 | NP_001192004.1 | 3 - 4 | 915 | 86 | |
GenBank mRNA | BC144865.1 | - | 3 - 4 | 603 | 9053413 | 86 |
D50055.1 | - | 3 - 4 | 598 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |