Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139227.4 | NP_631973.3 | 6 - 7 | 1242 | 105 | |
GenBank mRNA | AF455111.1 | - | 5 - 6 | 982 | 105 | |
AK046679.1 | - | 6 - 7 | 1243 | 105 | ||
BC035459.1 | - | 5 - 6 | 895 | 105 | ||
BC137846.1 | - | 6 - 7 | 1163 | 8860865 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |