Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 35 - 36 | 5133 | 67 | |
GenBank mRNA | AF093576.1 | - | 35 - 36 | 5041 | 67 | |
AK161449.1 | - | 6 - 7 | 774 | 67 | ||
BC150747.1 | - | 35 - 36 | 4998 | 9087658 | 67 | |
M10276.1 | - | 3 - 4 | 270 | 67 | ||
U87455.1 | - | 35 - 36 | 5113 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |