Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 50 - 51 | 7136 | 118 | |
GenBank mRNA | AF093576.1 | - | 50 - 51 | 7044 | 118 | |
AK086017.1 | - | 7 - 8 | 776 | 118 | ||
AK143823.1 | - | 14 - 15 | 1804 | 118 | ||
AK161449.1 | - | 20 - 21 | 4062 | 118 | ||
BC150747.1 | - | 50 - 51 | 7001 | 9087658 | 118 | |
U87455.1 | - | 50 - 51 | 7116 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |