Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpysl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009955.3 | NP_034085.2 | 4 - 5 | 705 | 70 | |
GenBank mRNA | BC062955.1 | - | 4 - 5 | 704 | 30535984 | 70 |
X87242.1 | - | 4 - 5 | 602 | 70 | ||
Y10339.1 | - | 4 - 5 | 495 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |