Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110162.1 | NP_001103632.1 | 10 - 11 | 1715 | 110 | |
NM_175384.4 | NP_780593.3 | 10 - 11 | 1418 | 110 | ||
GenBank mRNA | AK031313.1 | - | 6 - 7 | 860 | 110 | |
AK160223.1 | - | 10 - 11 | 1478 | 110 | ||
AK166952.1 | - | 10 - 11 | 1485 | 110 | ||
BC037136.1 | - | 10 - 11 | 1515 | 110 | ||
BC067009.1 | - | 10 - 11 | 1446 | 110 | ||
BC075682.1 | - | 10 - 11 | 1418 | 110 | ||
BC116298.1 | - | 6 - 7 | 860 | 40095010 | 110 | |
BC116299.1 | - | 6 - 7 | 860 | 40095011 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |