Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133857.3 | NP_598618.1 | 4 - 5 | 1052 | 71 | |
GenBank mRNA | AK030317.1 | - | 5 - 6 | 1136 | 71 | |
AK036876.1 | - | 4 - 5 | 1053 | 71 | ||
AK045953.1 | - | 4 - 5 | 1115 | 71 | ||
AK140453.1 | - | 5 - 6 | 1015 | 71 | ||
BC096383.1 | - | 4 - 5 | 1052 | 71 | ||
BC132339.1 | - | 3 - 4 | 785 | 40130616 | 71 | |
BC145707.1 | - | 3 - 4 | 785 | 40131010 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |