Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080815.1 | NP_001074284.1 | 5 - 6 | 537 | 82 | |
GenBank mRNA | AK050472.1 | - | 5 - 6 | 576 | 82 | |
AK133386.1 | - | 1 - 2 | 62 | 82 | ||
AK146552.1 | - | 5 - 6 | 539 | 82 | ||
AY255568.1 | - | 3 - 4 | 82 | |||
BC120671.1 | - | 4 - 5 | 441 | 40129594 | 82 | |
BC120673.1 | - | 4 - 5 | 441 | 40129596 | 82 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |