Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700002140.mm.1 |
TC0700000285.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002140.mm.1 |
NM_001190490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002140.mm.1 |
NM_001190490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002140.mm.1 |
NM_001190491.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002140.mm.1 |
NM_001190491.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002140.mm.1 |
NM_032418.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002140.mm.1 |
NM_032418.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002137.mm.1 |
TC0700000285.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002137.mm.1 |
NM_001190490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002137.mm.1 |
NM_001190490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002137.mm.1 |
NM_001190491.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002137.mm.1 |
NM_001190491.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002137.mm.1 |
NM_032418.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002137.mm.1 |
NM_032418.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002143.mm.1 |
TC0700000285.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002143.mm.1 |
NM_001190490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002143.mm.1 |
NM_001190490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002143.mm.1 |
NM_001190491.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002143.mm.1 |
NM_001190491.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002143.mm.1 |
NM_032418.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002143.mm.1 |
NM_032418.2 |
Refseq |