Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptger3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011196.2 | NP_035326.2 | 1 - 2 | 952 | 79 | |
GenBank mRNA | BC058742.1 | - | 1 - 2 | 952 | 6309451 | 79 |
D10204.1 | - | 1 - 2 | 941 | 79 | ||
D13321.1 | - | 1 - 2 | 941 | 79 | ||
D17406.1 | - | 1 - 2 | 941 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |