Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Extl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018788.3 | NP_061258.2 | 4 - 5 | 2941 | 66 | |
GenBank mRNA | AK137842.1 | - | 2 - 3 | 2018 | 66 | |
AK220344.1 | - | 2 - 3 | 2284 | 66 | ||
BC058526.1 | - | 2 - 3 | 2332 | 66 | ||
BC065073.1 | - | 2 - 3 | 2406 | 6834580 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |