Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp3cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015002.1 | - | 6 - 7 | 995 | 94 | |
AK133393.1 | - | 6 - 7 | 1016 | 94 | ||
BC046617.1 | - | 6 - 7 | 821 | 94 | ||
BC141079.1 | - | 6 - 7 | 836 | 9055691 | 94 | |
BC145347.1 | - | 5 - 6 | 711 | 94 | ||
M81475.1 | - | 6 - 7 | 989 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |