Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199003.1 | NP_001185932.1 | 8 - 9 | 860 | 103 | |
NM_011880.3 | NP_036010.2 | 8 - 9 | 860 | 103 | ||
GenBank mRNA | AF011360.1 | - | 8 - 9 | 705 | 103 | |
BC051133.1 | - | 8 - 9 | 815 | 6588901 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |