Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700010454.mm.1 |
TC0700001185.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010454.mm.1 |
NM_001277954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010454.mm.1 |
NM_001277955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010454.mm.1 |
NM_017400.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010452.mm.1 |
TC0700001185.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010452.mm.1 |
NM_001277954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010452.mm.1 |
NM_001277955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010452.mm.1 |
NM_017400.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010453.mm.1 |
TC0700001185.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010453.mm.1 |
NM_001277954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010453.mm.1 |
NM_001277955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010453.mm.1 |
NM_017400.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010455.mm.1 |
TC0700001185.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700010455.mm.1 |
NM_001277954.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010455.mm.1 |
NM_001277955.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700010455.mm.1 |
NM_017400.7 |
Refseq |