Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nexn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199465.2 | NP_955759.2 | 6 - 7 | 646 | 131 | |
GenBank mRNA | AK136102.1 | - | 6 - 7 | 574 | 131 | |
AK162232.1 | - | 1 - 2 | 87 | 131 | ||
AK163414.1 | - | 6 - 7 | 646 | 131 | ||
AK163848.1 | - | 5 - 6 | 560 | 131 | ||
BC052878.1 | - | 6 - 7 | 593 | 30046316 | 131 | |
HQ179982.1 | - | 6 - 7 | 680 | 131 | ||
HQ179983.1 | - | 5 - 6 | 638 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |