Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrriq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028938.2 | NP_083214.2 | 5 - 6 | 1104 | 161 | |
GenBank mRNA | AK015333.1 | - | 5 - 6 | 1001 | 161 | |
AK016634.1 | - | 5 - 6 | 999 | 161 | ||
AK132656.1 | - | 5 - 6 | 1069 | 161 | ||
BC112424.1 | - | 5 - 6 | 961 | 40062088 | 161 | |
BC113130.1 | - | 5 - 6 | 961 | 40062093 | 161 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |