Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026474.5 | NP_080750.1 | 5 - 6 | 462 | 128 | |
GenBank mRNA | AK010711.1 | - | 4 - 5 | 431 | 128 | |
AK010830.1 | - | 5 - 6 | 379 | 128 | ||
AK020433.1 | - | 5 - 6 | 381 | 128 | ||
AK045153.1 | - | 5 - 6 | 401 | 128 | ||
AK082328.1 | - | 5 - 6 | 464 | 128 | ||
AK141416.1 | - | 398 | 128 | |||
AK148838.1 | - | 5 - 6 | 376 | 128 | ||
BC009167.1 | - | 5 - 6 | 379 | 2811924 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |